Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant RNF8 (from E. coli), Unconjugated
Human Recombinant RNF8 (from E. coli), Unconjugated
Supplier:  BIORTUS USA
Certificates
undefined
Human Recombinant RNF8 (from E. coli), Unconjugated
Supplier:  BIORTUS USA

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt O76064
  • Protein/Peptide Name:
    RNF8
  • Purity:
    80%
  • Sequence:
    G2-F485
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications