Human Recombinant RNF181 (from E. coli), His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9P0P0
- Protein/Peptide Name:RNF181
- Purity:90%
- Sequence:A2-T153
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice