Human Recombinant RFPL3 (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt O75679
- Protein/Peptide Name:RFPL3
- Purity:90%
- Sequence:K2-K317
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 150 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together