Human Recombinant RBCK1 (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9BYM8
- Protein/Peptide Name:RBCK1
- Purity:80%
- Sequence:D2-H510
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice