Human Recombinant PPIL2 (from E. coli), Unconjugated
Supplier: Biortus
About this item
Delivery information: Frozen liquid on dry ice.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Delivery information: Frozen liquid on dry ice.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 76792-590
- 76793-906
- 76795-288
- 76792-612
- 76794-084
- Protein/Peptide Name:
- PPIL2
- PPIL2
- PPIL2
- PPIL2
- PPIL2
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- UniProt Q13356
- UniProt Q13356
- UniProt Q13356
- UniProt Q13356
- UniProt Q13356
- Sequence:
- G2-W520
- G2-W520
- G2-W520
- G2-W520
- G2-W520
- Purity:
- 85%
- 85%
- 85%
- 85%
- 0.85
- Formulation:
- 50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- 50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- 50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- 50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- 50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- Shipping Temperature:
- Frozen liquid with dry ice
- Frozen liquid with dry ice
- Frozen liquid with dry ice
- Frozen liquid with dry ice
- Frozen liquid with dry ice
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: BP08543-00A-1SItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76792-590Protein/Peptide NamePPIL2Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDUniProt Q13356SequenceG2-W520Purity85%Formulation50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerolShipping TemperatureFrozen liquid with dry ice - Supplier Number: BP08543-00A-1BItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76793-906Protein/Peptide NamePPIL2Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDUniProt Q13356SequenceG2-W520Purity85%Formulation50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerolShipping TemperatureFrozen liquid with dry ice - Supplier Number: BP08543-00A-2BItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76795-288Protein/Peptide NamePPIL2Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDUniProt Q13356SequenceG2-W520Purity85%Formulation50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerolShipping TemperatureFrozen liquid with dry ice - Supplier Number: BP08543-00A-5BItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76792-612Protein/Peptide NamePPIL2Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDUniProt Q13356SequenceG2-W520Purity85%Formulation50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerolShipping TemperatureFrozen liquid with dry ice - Supplier Number: BP08543-00A-1QItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76794-084Protein/Peptide NamePPIL2Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDUniProt Q13356SequenceG2-W520Purity0.85Formulation50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerolShipping TemperatureFrozen liquid with dry ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



