Human Recombinant PELI2 (from E. coli), Unconjugated
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9HAT8
- Protein/Peptide Name:PELI2
- Purity:85%
- Sequence:F2-D420
- Formulation:50 mM Tris-HCl (pH 7.5), 250 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice