Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MDM4 (from E. coli), Unconjugated
Human Recombinant MDM4 (from E. coli), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant MDM4 (from E. coli), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt O15151
  • Protein Synonyms:
    p53-binding protein Mdm4
  • Protein/Peptide Name:
    MDM4
  • Purity:
    85%
  • Sequence:
    T2-A490
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together