Human Recombinant BIRC7 (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q96CA5
- Protein/Peptide Name:BIRC7
- Purity:85%
- Sequence:G2-S298
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together