Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IVNS1ABP (from Insect cells), Unconjugated
 :  Biortus
 
undefined
Human Recombinant IVNS1ABP (from Insect cells), Unconjugated
 :  Biortus

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q9Y6Y0
  • Protein/Peptide Name:
    IVNS1ABP
  • Purity:
    80%
  • Sequence:
    I2-F642 end
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

 

Frequently Bought Together