Human Recombinant HLTF (from E. coli), His
: Frozen liquid on dry ice.
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q14527
- Protein Synonyms:RING finger protein 80
- Protein/Peptide Name:HLTF
- Purity:90%
- Sequence:V58-A174
- Formulation:50 mM Tris-HCl pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice