Human Recombinant HACE1 (from E. coli), His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8IYU2
- Protein/Peptide Name:HACE1
- Purity:85%
- Sequence:E2-A909
- Formulation:50 mM Tris-HCl pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together