Human Recombinant HACE1 (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8IYU2
- Protein/Peptide Name:HACE1
- Purity:80%
- Sequence:E2-A909
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice