Human Recombinant DTX3L (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8TDB6
- Protein/Peptide Name:DTX3L
- Purity:80%
- Sequence:A2-E740
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 150 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together