Human Recombinant DTX3 (from E. coli), Unconjugated
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8N9I9
- Protein Synonyms:RING finger protein 154
- Protein/Peptide Name:DTX3
- Purity:80%
- Sequence:S2-D347
- Formulation:50 mM Tris-HCl (pH7.5), 150 mM NaCl, 1 mM DTT, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice