Human Recombinant WDTC1/DDB1 (from Insect cells), His/StrepII
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/StrepII
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q8N5D0/Q16531
- Protein Synonyms:WD and tetratricopeptide repeats protein 1
- Protein/Peptide Name:WDTC1/DDB1
- Purity:90%
- Sequence:A2-S677 end/M1-H1140 end
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together