Human Recombinant Gene Name: SLC40A1 Protein (from Mammalian cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt Q9NP59
- Protein Synonyms:SLC
- Protein/Peptide Name:SLC40A1
- Purity:95%
- Sequence:M1-V571 end
- Formulation:20 mM HEPES pH 7.5, 100 mM NaCl, 0.03% (w/v) DDM, and 0.003% (w/v) CHS, Protease inhibitor (Biortus).
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together