Human Recombinant Gene Name: SLC6A9 Protein (from Mammalian cells), His/eGFP
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/eGFP
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt P48067
- Protein Synonyms:SLC
- Protein/Peptide Name:SLC6A9
- Purity:95%
- Sequence:M1-I706 end, L153A, S297A, I368A, C633A, Δ1–90, Δ240–256, Δ685–706
- Formulation:50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.003% (w/v) LMNG, 0.0003% (w/v) CHS
- Shipping Temperature:Frozen liquid with dry ice