Human Recombinant SLC12A2 Protein (from Mammalian cells), His/StrepII/eGFP
: Frozen liquid on dry ice.
- Conjugation:His/StrepII/eGFP
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt P55011
- Protein Synonyms:SLC
- Protein/Peptide Name:SLC12A2
- Purity:80%
- Sequence:M1-S1212 end
- Formulation:20 mM Tris-HCl pH 8.0, 200 mM NaCl, 200 mM KCl, 0.01% (w/v) GDN
- Shipping Temperature:Frozen liquid with dry ice