SARS-CoV-2 Recombinant S Protein (from Mammalian cells), Strep/His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Strep/His
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTC2
- Protein/Peptide Name:S
- Purity:80%
- Sequence:M1-Q1208, K986P, V987P
- Formulation:20 mM Tris-HCl (pH 7.5), 150 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together