SARS-CoV-2 Recombinant S Protein (from Insect cells), His
: Frozen liquid on dry ice.
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTC2
- Protein/Peptide Name:S
- Purity:90%
- Sequence:R319-F541
- Formulation:20 mM HEPES (pH 7.5), 150 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice