SARS-CoV-2 Recombinant rep Protein (from E. coli), His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTD1
- Protein/Peptide Name:rep
- Purity:95%
- Sequence:Q1092-E1202
- Formulation:20 mM HEPES (pH 8.0), 150 mM NaCl, 2 mM TCEP
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together