SARS-CoV-2 Recombinant rep Protein (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTD1
- Protein/Peptide Name:rep
- Purity:95%
- Sequence:S1-N298/ A1-Q139
- Formulation:50 mM Tris-HCl (pH 8.0), 500 mM NaCl, 5% glycerol, 2 mM MgSO4
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together