SARS-CoV-2 Recombinant NCP-N Protein (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTC9
- Protein/Peptide Name:NCP-N
- Purity:85%
- Sequence:N269-K369
- Formulation:10 mM Tris-HCl (pH 8.0), 150 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together