Human Recombinant ACE2 Protein (from Insect cells), His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9BYF1
- Protein/Peptide Name:ACE2
- Purity:90%
- Sequence:S19-D615
- Formulation:20 mM HEPES (pH 7.5), 300 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together