Human Recombinant NRAS Protein (from E. coli), biotinylated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P01111
- Protein Synonyms:Ras|GTPase NRas
- Protein/Peptide Name:NRAS
- Purity:95%
- Sequence:M1-N172, G13D
- Formulation:20 mM HEPES (pH 7.5), 1 mM MgCl₂, 150 mM NaCl, 0.5 mM TCEP.
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together