Human Recombinant KRAS Protein (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P01116
- Protein Synonyms:GTPase KRas|Ras
- Protein/Peptide Name:KRAS
- Purity:95%
- Sequence:M1-K169
- Formulation:50 mM HEPES (pH 7.4), 5 mM MgCl₂, 150 mM NaCl, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice