Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant KRAS Protein (from E. coli), biotinylated
 
undefined
Human Recombinant KRAS Protein (from E. coli), biotinylated

 

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt P01116
  • Protein Synonyms:
    GTPase KRas|Ras
  • Protein/Peptide Name:
    KRAS
  • Purity:
    95%
  • Sequence:
    M1-M188, Q61R
  • Formulation:
    20 mM HEPES (pH 7.5), 1 mM MgCl₂, 150 mM NaCl, 0.5 mM TCEP.
  • Shipping Temperature:
    Frozen liquid with dry ice