Human Recombinant ATP2A2 Protein (from Mammalian cells), His/StrepII/eGFP
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/StrepII/eGFP
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt P16615
- Protein Synonyms:SERCA2
- Protein/Peptide Name:ATP2A2
- Purity:90%
- Sequence:M1-S1042 end
- Formulation:50 mM HEPES pH 7.5, 100 mM KCl , 1 mM DTT, 1 mM CaCl₂, 1 mM MgCl₂, 0.001% (w/v) LMNG.
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together