Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant ZDHHC20 Protein (from Insect cells), Unconjugated
Human Recombinant ZDHHC20 Protein (from Insect cells), Unconjugated
Supplier:  BIORTUS USA
Certificates
undefined
Human Recombinant ZDHHC20 Protein (from Insect cells), Unconjugated
Supplier:  BIORTUS USA

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q5W0Z9
  • Protein Synonyms:
    DHHC20
  • Protein/Peptide Name:
    ZDHHC20
  • Purity:
    85%
  • Sequence:
    M1-N365 end
  • Formulation:
    50 mM HEPES pH 7.5, 150 mM NaCl, 2 mM TCEP, 0.025% (w/v) DDM.
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications