Human Recombinant SIGMAR1 Protein (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q99720
- Protein/Peptide Name:SIGMAR1
- Purity:90%
- Sequence:M1-P223 end
- Formulation:20 mM HEPES pH 7.5, 100 mM NaCl, 0.01% (w/v) LMNG, 0.001% (w/v) CHS, 1 μM dextromethorphan.
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together