Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant SCD Protein (from Insect cells), His
Human Recombinant SCD Protein (from Insect cells), His
Supplier:  BIORTUS USA
Certificates
undefined
Human Recombinant SCD Protein (from Insect cells), His
Supplier:  BIORTUS USA

Specifications

  • Conjugation:
    His
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt O00767
  • Protein/Peptide Name:
    SCD
  • Purity:
    95%
  • Sequence:
    I46-G359, K60A, K62A, E63A
  • Formulation:
    20 mM HEPES pH 7.5, 150 mM NaCl, 0.02% (w/v) DDM, 0.002% (w/v) CHS.
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications