Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant RPS6KA6 Protein (from Insect cells), GST
 :  Biortus
 
undefined
Human Recombinant RPS6KA6 Protein (from Insect cells), GST
 :  Biortus

 

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q9UK32
  • Protein Synonyms:
    Ribosomal protein S6 kinase alpha-6
  • Protein/Peptide Name:
    RPS6KA6
  • Purity:
    90%
  • Sequence:
    M1-L745
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 0.05% Brij35, 10% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

 

Frequently Bought Together