Human Recombinant RPS6KA2 Protein (from Insect cells), GST
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q15349
- Protein Synonyms:p90-RSK 2
- Protein/Peptide Name:RPS6KA2
- Purity:90%
- Sequence:M1-L733
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl , 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together