Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PRKG2 Protein (from Insect cells), GST
Human Recombinant PRKG2 Protein (from Insect cells), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant PRKG2 Protein (from Insect cells), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q13237
  • Protein Synonyms:
    cGMP|cGK2
  • Protein/Peptide Name:
    PRKG2
  • Purity:
    90%
  • Sequence:
    M1-F762
  • Formulation:
    20 mM Tris-HCl (pH 7.5), 150 mM NaCl, 1 mM DTT, 5% glycerol.
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together