Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PRKCB Protein (from Insect cells), His/GST
Human Recombinant PRKCB Protein (from Insect cells), His/GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant PRKCB Protein (from Insect cells), His/GST
Supplier:  Biortus

Specifications

  • Conjugation:
    His/GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt P05771
  • Protein/Peptide Name:
    PRKCB
  • Purity:
    90%
  • Sequence:
    A2-S673
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 20% glycerol, 5 mM DTT, 0.1 M Trehalose
  • Shipping Temperature:
    Liquid. It is shipped out with dry ice

Specifications

Related Information

Frequently Bought Together