Human Recombinant PIM2 Protein (from E. coli), Unconjugated
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q9P1W9
- Protein/Peptide Name:PIM2
- Purity:90%
- Sequence:M1-P311
- Formulation:50 mM HEPES (pH 7.5), 300 mM NaCl, 20% glycerol, 0.5 mM TCEP
- Shipping Temperature:Frozen liquid with dry ice