Human Recombinant PIM1 Protein (from E. coli), Unconjugated
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P11309
- Protein/Peptide Name:PIM1
- Purity:90%
- Sequence:A14-K313
- Formulation:50 mM HEPES (pH 7.5), 150 mM NaCl, 5% glycerol.
- Shipping Temperature:Frozen liquid with dry ice