Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PASK Protein (from Insect cells), GST
Human Recombinant PASK Protein (from Insect cells), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant PASK Protein (from Insect cells), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q96RG2
  • Protein/Peptide Name:
    PASK
  • Purity:
    90%
  • Sequence:
    S949-S1323
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 0.05% Brij35, 10% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together