Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MAPK14 Protein (from Insect cells), GST
Human Recombinant MAPK14 Protein (from Insect cells), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant MAPK14 Protein (from Insect cells), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q16539
  • Protein/Peptide Name:
    MAPK14
  • Purity:
    90%
  • Sequence:
    S2-S360
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together