Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant STK26 Protein (from Insect cells), GST
Human Recombinant STK26 Protein (from Insect cells), GST
Supplier:  BIORTUS USA
Certificates
undefined
Human Recombinant STK26 Protein (from Insect cells), GST
Supplier:  BIORTUS USA

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q9P289
  • Protein Synonyms:
    MST-4
  • Protein/Peptide Name:
    STK26
  • Purity:
    90%
  • Sequence:
    M1-P416
  • Formulation:
    50 mM Tris (pH 7.5), 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton® X–100, 2 mM DTT, 10% glycerol
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications