Human Recombinant STK24 Protein (from Insect cells), GST
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9Y6E0
- Protein/Peptide Name:STK24
- Purity:90%
- Sequence:M1-H443
- Formulation:50 mM Tris (pH 7.5), 200 mM NaCl, 0.05% Brij35, 1 mM DTT, 10% Glycerol
- Shipping Temperature:Frozen liquid with dry ice