Human Recombinant STK3 Protein (from Insect cells), GST
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q13188
- Protein Synonyms:MST-2
- Protein/Peptide Name:STK3
- Purity:90%
- Sequence:M1-F491
- Formulation:50 mM Tris (pH 7.5), 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton® X–100, 2 mM DTT, 10% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together