Human Recombinant MET Protein (from Insect cells), His/GST
: Frozen liquid on dry ice.
- Conjugation:His/GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P08581
- Protein/Peptide Name:MET
- Purity:90%
- Sequence:K956-S1390
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice