Human Recombinant LYN Protein (from Insect cells), GST
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P07948
- Protein/Peptide Name:LYN
- Purity:90%
- Sequence:G2-P512
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 20% glycerol, 5 mM DTT, 100 mM Trehalose
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together