Human Recombinant IRAK4 Protein (from Insect cells), His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9NWZ3
- Protein/Peptide Name:IRAK4
- Purity:90%
- Sequence:E154-S460
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice