Human Recombinant GSK3A Protein (from Insect cells), GST
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P49840
- Protein/Peptide Name:GSK3A
- Purity:90%
- Sequence:S2-S483
- Shipping Temperature:Liquid. It is shipped out with dry ice