Human Recombinant FGR Protein (from Insect cells), GST
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P09769
- Protein Synonyms:Tyrosine-protein kinase Fgr
- Protein/Peptide Name:FGR
- Purity:90%
- Sequence:G2-T529
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 20% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice