Human Recombinant FES Protein (from E. coli), His
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P07332
- Protein/Peptide Name:FES
- Purity:90%
- Sequence:I448-R822
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice