Human Recombinant EPHA3 Protein (from Insect cells), GST
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P29320
- Protein Synonyms:HEK|EK4|hEK4
- Protein/Peptide Name:EPHA3
- Purity:90%
- Sequence:K579-V983
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5 mM DTT, 20% glycerol, 0.1 mM Trehalose
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together