Human Recombinant DTL/DDB1 (from Insect cells), His/StrepII
Supplier: BIORTUS USA
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/StrepII
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9NZJ0/Q16531
- Protein Synonyms:DDB1- and CUL4-associated factor 2
- Protein/Peptide Name:DTL/DDB1
- Purity:90%
- Sequence:L2-L730 end/M1-H1140 end
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice